Assay Details
Target Gene Details
Entrez Gene ID: | 71045 |
Gene Name: | RIKEN cDNA 4933406I18 gene |
Gene Aliases: |
4933403D24Rik |
Location: |
Chr.7:114315479-114415161 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4933406I18Rik | NR_029437.1 | 1 | 223 | |
NR_029438.1 | 1 | 223 | ||
AK016630.1 | 1 | 84 | ||
AK016693.1 | 1 | 229 |
Target Gene Details
Entrez Gene ID: | 18576 |
Gene Name: | phosphodiesterase 3B, cGMP-inhibited |
Gene Aliases: |
9830102A01Rik, AI847709 |
Location: |
Chr.7:114414700-114537938 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde3b |