Assay Details
Target Gene Details
Entrez Gene ID: | 233529 |
Gene Name: | potassium channel tetramerisation domain containing 14 |
Gene Aliases: |
AI449310, D7Ertd760e |
Location: |
Chr.7:97436380-97459557 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kctd14 | NM_001010826.3 | 1 | 126 | NP_001010826.2 |
NM_001012434.3 | 1 | 126 | NP_001012436.1 | |
NM_001136235.1 | NP_001129707.1 | |||
AK145279.1 | 1 | 127 | BAE26341.1 | |
AY899809.1 | 1 | 126 | AAW88505.1 | |
BG088171.2 | ||||
BY735425.1 | 1 | 127 | ||
CK791017.1 |
More Information
Set Membership: |
Intragenic Exonic |