Assay Details
Target Gene Details
Entrez Gene ID: | 384724 |
Gene Name: | cytochrome P450, family 2, subfamily t, polypeptide 4 |
Gene Aliases: |
Gm1448, Gm1450 |
Location: |
Chr.7:27153714-27158973 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp2t4 |
Target Gene Details
Entrez Gene ID: | 112406 |
Gene Name: | egl-9 family hypoxia-inducible factor 2 |
Gene Aliases: |
0610011A13Rik, C85656, Hif-p4h-1, Ier4, Phd1, SM-20 |
Location: |
Chr.7:27158658-27166802 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egln2 | NM_053208.4 | 6 | 1921 | NP_444438.2 |
AF340231.1 | 5 | 1663 | AAK37525.1 | |
AF453879.1 | 6 | 1843 | AAL65166.1 | |
AJ310547.1 | 6 | 1892 | CAC42516.1 | |
AK075582.1 | 6 | 1873 | BAC35835.1 | |
AK152056.1 | 6 | 1882 | BAE30912.1 | |
AK152223.1 | 6 | 1879 | BAE31050.1 | |
AK159206.1 | 6 | 1874 | BAE34897.1 | |
AK166661.1 | 7 | 1787 | ||
AK167477.1 | 6 | 1874 | ||
BC023299.1 | 6 | 1855 | AAH23299.2 | |
BC051436.1 | 5 | 1639 | AAH51436.1 | |
BC086764.1 | 6 | 1860 | AAH86764.1 | |
BE993075.1 | 1 | 204 |