Assay Details
Target Gene Details
Entrez Gene ID: | 13626 |
Gene Name: | embryonic ectoderm development |
Gene Aliases: |
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk |
Location: |
Chr.7:89954654-89981419 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eed | NM_021876.3 | 12 | 1722 | NP_068676.1 |
AK077664.1 | 12 | 1307 | BAC36938.1 | |
AK087924.1 | 12 | 1283 | ||
AK131976.1 | 12 | 1289 | BAE20916.1 | |
AK133890.1 | 12 | 1283 | BAE21915.1 | |
AK213110.1 | 1 | 40 | ||
BC012966.1 | 12 | 1721 | AAH12966.1 | |
CJ200028.1 | 1 | 56 | ||
U78103.1 | 12 | 1724 | AAB38319.1 | |
U97675.1 | 12 | 1729 | AAC53302.1 |
More Information
Set Membership: |
Intragenic Exonic |