Assay Details
Target Gene Details
Entrez Gene ID: | 67143 |
Gene Name: | IKAROS family zinc finger 5 |
Gene Aliases: |
2610034F18Rik, PEGASUS, Zfpn1a5 |
Location: |
Chr.7:131388649-131410506 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ikzf5 | NM_175115.4 | 4 | 3081 | NP_780324.2 |
AK009564.1 | 1 | 457 | ||
AK148824.1 | 1 | 686 | ||
AK153883.1 | 4 | 3096 | BAE32231.1 | |
BC048183.1 | 4 | 3081 | AAH48183.1 | |
BC058537.1 | 4 | 3081 | AAH58537.1 |
Target Gene Details
Entrez Gene ID: | 214580 |
Gene Name: | phosphoseryl-tRNA kinase |
Gene Aliases: |
5430423O14Rik, 5730458D16Rik, Gm1099, R75284 |
Location: |
Chr.7:131371113-131392768 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pstk | AK028520.1 | 1 | 770 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |