Assay Details
Target Gene Details
Entrez Gene ID: | 269951 |
Gene Name: | isocitrate dehydrogenase 2 (NADP+), mitochondrial |
Gene Aliases: |
E430004F23, IDPm, Idh-2 |
Location: |
Chr.7:80094846-80115387 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idh2 | NM_173011.2 | 11 | 1408 | NP_766599.2 |
AF212319.1 | 11 | 1379 | AAG43538.1 | |
AK088110.1 | 11 | 1380 | BAC40149.1 | |
AK145753.1 | 11 | 1353 | BAE26628.1 | |
AK146564.1 | 11 | 1494 | ||
AK161640.1 | 11 | 1392 | BAE36505.1 | |
AK169395.1 | 11 | 1405 | BAE41142.1 | |
AK179419.1 | 2 | 111 | ||
AK209654.1 | 2 | 111 | ||
AK212737.1 | 2 | 111 | ||
BC060030.1 | 11 | 1396 | AAH60030.1 | |
BQ747924.1 | 2 | 158 |
More Information
Set Membership: |
Intragenic Exonic |