Assay Details
Target Gene Details
Entrez Gene ID: | 20349 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E |
Gene Aliases: |
6430702L12, AA408817, Semah, mKIAA0331 |
Location: |
Chr.5:14025276-14256689 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sema3e | NM_011348.2 | 17 | 3958 | NP_035478.2 |
AK129116.1 | 12 | 2723 | BAC97926.1 | |
AK141295.1 | 16 | 4681 | ||
Z93947.1 | 17 | 3936 | CAB07987.1 | |
Z93948.1 | CAB07988.1 |
More Information
Set Membership: |
Intragenic Exonic |