Assay Details
Target Gene Details
Entrez Gene ID: | 67602 |
Gene Name: | NECAP endocytosis associated 1 |
Gene Aliases: |
1200016B17Rik, AI747569, mFLJ00061 |
Location: |
Chr.6:122874557-122888941 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Necap1 | NM_026267.2 | 8 | 2124 | NP_080543.2 |
AK004805.1 | 8 | 2125 | BAB23577.2 | |
AK005263.1 | 8 | 2115 | BAB23915.2 | |
AK081598.1 | 8 | 2125 | BAC38266.1 | |
AK159237.1 | 1 | 1234 | ||
AK168500.1 | 8 | 2125 | BAE40385.1 | |
AK220236.1 | 8 | 2110 | BAD90161.1 | |
BC011466.1 | 8 | 2123 |
More Information
Set Membership: |
Intragenic Exonic |