Assay Details
Target Gene Details
Entrez Gene ID: | 232748 |
Gene Name: | TRPM8 channel-associated factor 2 |
Gene Aliases: |
Fam115c |
Location: |
Chr.6:42623043-42645041 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcaf2 | NM_146174.1 | 3 | 972 | NP_666286.1 |
AK131169.1 | 3 | 1185 | BAD21419.1 | |
BC008555.1 | 3 | 989 | AAH08555.1 | |
BC011487.1 | 3 | 972 | AAH11487.1 |