Assay Details
Target Gene Details
Entrez Gene ID: | 21333 |
Gene Name: | tachykinin 1 |
Gene Aliases: |
4930528L02Rik, NK-1, NK1, Nkna, PPT-A, PPTA, SP |
Location: |
Chr.6:7554776-7562978 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tac1 | NM_009311.2 | 7 | 535 | NP_033337.1 |
BC117081.1 | 7 | 510 | ||
BC119122.1 | 7 | 510 | ||
BC144737.1 | 6 | 465 | ||
D17584.1 | 7 | 535 | BAA04508.1 |