Assay Details
Target Gene Details
Entrez Gene ID: | 243653 |
Gene Name: | C-type lectin domain family 1, member a |
Gene Aliases: |
5930406N14Rik |
Location: |
Chr.6:129423358-129452000 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec1a | NM_175526.3 | 6 | 1335 | NP_780735.2 |
AK031121.1 | 6 | 1157 | BAC27264.1 | |
AK036846.1 | 6 | 1138 | BAC29605.1 | |
AK049608.1 | 6 | 1082 | BAC33840.1 | |
AK165920.1 | 6 | 1195 | ||
BC052840.1 | 6 | 1194 | AAH52840.1 |
Target Gene Details
Entrez Gene ID: | 232414 |
Gene Name: | C-type lectin domain family 9, member a |
Gene Aliases: |
9830005G06Rik, DNGR-1 |
Location: |
Chr.6:129408862-129501701 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec9a |