Assay Details
Target Gene Details
Entrez Gene ID: | 109978 |
Gene Name: | ADP-ribosyltransferase 4 |
Gene Aliases: |
4432404K01Rik, ARTC4, DO, DOK1 |
Location: |
Chr.6:136848451-136857600 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Art4 | NM_026639.2 | 1 | 368 | NP_080915.1 |
AK004744.1 | 1 | 322 | BAB23525.1 | |
AK019467.1 | 1 | 369 | BAB31738.1 | |
BC046306.1 | 1 | 289 | AAH46306.1 |
More Information
Set Membership: |
Intragenic Exonic |