Assay Details
Target Gene Details
Entrez Gene ID: | 170676 |
Gene Name: | paternally expressed 10 |
Gene Aliases: |
AA407948, Edr, HB-1, MEF3L, Mar2, Mart2, MyEF-3 |
Location: |
Chr.6:4747306-4760517 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Peg10 | NM_001040611.1 | 1 | 88 | NP_001035701.1 |
NM_130877.2 | 1 | 88 | NP_570947.2 | |
AB091827.1 | BAC77244.1 | |||
AJ006464.1 | ||||
CJ127881.1 | 1 | 88 |