Assay Details
Target Gene Details
Entrez Gene ID: | 22628 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide |
Gene Aliases: |
14-3-3gamma, D7Bwg1348e |
Location: |
Chr.5:135908379-135934641 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ywhag | NM_018871.3 | 2 | 2501 | NP_061359.2 |
AK088847.1 | 2 | 2414 | BAC40609.1 | |
AK153307.1 | 2 | 2460 | BAE31888.1 | |
AK164356.1 | 2 | 2452 | BAE37756.1 | |
AK164696.1 | 1 | 445 | BAE37881.1 | |
AK188965.1 | ||||
AK201168.1 | 1 | 85 | ||
BC008129.1 | 2 | 2384 | AAH08129.1 |
More Information
Set Membership: |
Intragenic Exonic |