Assay Details
Target Gene Details
Entrez Gene ID: | 18575 |
Gene Name: | phosphodiesterase 1C |
Gene Aliases: |
- |
Location: |
Chr.6:56069798-56652516 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde1c | NM_001159953.1 | NP_001153425.1 | ||
NM_001159955.1 | 19 | 2809 | NP_001153427.1 | |
NM_001159956.1 | 17 | 2666 | NP_001153428.1 | |
NM_011054.4 | 19 | 3224 | NP_035184.1 | |
AK030423.1 | BAC26956.1 | |||
AK082497.1 | 18 | 3032 | ||
AK133243.1 | ||||
L76947.1 | AAC37702.1 |
More Information
Set Membership: |
Intragenic Exonic |