Assay Details
Target Gene Details
Entrez Gene ID: | 232440 |
Gene Name: | H2A histone family, member J |
Gene Aliases: |
E130307C13 |
Location: |
Chr.6:136808248-136810074 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2afj | NM_177688.4 | 1 | 1522 | NP_808356.1 |
AK053755.1 | 1 | 1447 | BAC35508.1 |
More Information
Set Membership: |
Intragenic Exonic |