Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp2 | NM_001286607.1 | 17 | 4312 | NP_001273536.1 |
NM_053242.4 | 19 | 4712 | NP_444472.2 | |
NM_212435.1 | 17 | 4375 | NP_997600.1 | |
AK031751.1 | 1 | 1983 | ||
AK042719.1 | 1 | 623 | ||
AK053022.1 | 1 | 395 | ||
BC062926.1 | 17 | 4375 | AAH62926.1 |