Assay Details
Target Gene Details
Entrez Gene ID: | 108099 |
Gene Name: | protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Gene Aliases: |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Location: |
Chr.5:24862735-25101906 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkag2 | NM_001170555.1 | 12 | 1492 | NP_001164026.1 |
NM_001170556.1 | 12 | 1489 | NP_001164027.1 | |
NM_145401.2 | 16 | 2571 | NP_663376.2 | |
AK032238.1 | 14 | 2083 | BAC27775.1 | |
AK034765.1 | 10 | 1157 | BAC28827.1 | |
AK154296.1 | 16 | 2572 | BAE32497.1 | |
AK165834.1 | 12 | 1431 | BAE38403.1 | |
AY348864.1 | 12 | 1422 | AAQ55224.1 | |
BC015283.1 | 16 | 2359 | AAH15283.1 |
More Information
Set Membership: |
Intragenic Exonic |