Assay Details
Target Gene Details
Entrez Gene ID: | 13841 |
Gene Name: | Eph receptor A7 |
Gene Aliases: |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
Location: |
Chr.4:28813044-28967511 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha7 | NM_001122889.1 | 5 | 1341 | NP_001116361.1 |
NM_001290434.1 | 5 | 1355 | NP_001277363.1 | |
NM_010141.4 | 5 | 1355 | NP_034271.3 | |
AK030480.1 | 5 | 1322 | BAC26982.1 | |
AK032973.1 | 5 | 1160 | BAE43283.1 | |
AK033903.1 | 5 | 1356 | BAC28509.1 | |
AK049848.1 | 5 | 1322 | ||
BC026153.1 | 5 | 1247 | AAH26153.1 | |
X79082.1 | 5 | 1334 | CAA55687.1 | |
X79083.1 | 5 | 1334 | CAA55688.1 | |
X79084.1 | 5 | 1334 | CAA55689.1 |
More Information
Set Membership: |
Intragenic Exonic |