Assay Details
Target Gene Details
Entrez Gene ID: | 140904 |
Gene Name: | calneuron 1 |
Gene Aliases: |
9630012C17Rik, Cabp8, MNCb-0849 |
Location: |
Chr.5:130369356-130846325 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Caln1 | NM_021371.2 | 6 | 1887 | NP_067346.1 |
NM_181045.1 | 7 | 1587 | NP_851388.1 | |
AB041539.1 | 6 | 1878 | BAA95024.1 | |
AF282251.1 | 7 | 1569 | AAK15156.1 | |
AK081011.1 | 6 | 1321 | BAC38115.1 | |
AK158641.1 | 6 | 1249 | BAE34592.1 | |
AK158892.1 | 6 | 1859 | BAE34716.1 | |
AK163648.1 | 6 | 1350 | BAE37437.1 | |
BC057129.1 | 6 | 1866 | AAH57129.1 | |
BC058963.1 | 6 | 1866 | AAH58963.1 |