Assay Details
Target Gene Details
Entrez Gene ID: | 23833 |
Gene Name: | CD52 antigen |
Gene Aliases: |
AI463198, B7, B7-Ag, CAMPATH-1, CLS1, MB7 |
Location: |
Chr.4:134093538-134095073 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd52 | NM_013706.2 | 2 | 448 | NP_038734.1 |
AK088161.1 | 2 | 415 | BAC40183.1 | |
AK155728.1 | 2 | 449 | BAE33405.1 | |
BC027495.1 | 2 | 398 | AAH27495.1 | |
M55561.1 | 2 | 437 | AAA39889.1 | |
X56745.1 | 1 | 192 | ||
X76696.1 | 2 | 424 | CAA54125.1 |