Assay Details
Target Gene Details
Entrez Gene ID: | 230316 |
Gene Name: | multiple EGF-like-domains 9 |
Gene Aliases: |
4933405H16Rik, 9630025C07, Egfl5, mKIAA0818 |
Location: |
Chr.4:70431927-70534928 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Megf9 | NM_172694.2 | 5 | 1231 | NP_766282.1 |
AK035987.1 | 5 | 1232 | BAC29269.1 | |
AK039319.1 | 5 | 1232 | BAC30317.1 | |
BC088994.1 | 5 | 1140 | ||
BC138128.1 | 5 | 1170 | ||
BC138129.1 | 5 | 1170 |
More Information
Set Membership: |
Intragenic Exonic |