Assay Details
Target Gene Details
Entrez Gene ID: | 12909 |
Gene Name: | calcitonin gene-related peptide-receptor component protein |
Gene Aliases: |
AL022669 |
Location: |
Chr.5:130029306-130060787 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crcp | NM_007761.2 | 6 | 564 | NP_031787.1 |
AF028242.1 | 6 | 554 | AAB84046.1 | |
AF118271.1 | 6 | 564 | AAD17244.1 | |
AK075587.1 | 6 | 559 | ||
AK178797.1 | 1 | 54 | ||
AK208487.1 | 1 | 54 | ||
AK211219.1 | 1 | 54 | ||
BC012676.1 | 6 | 498 | AAH12676.1 |
More Information
Set Membership: |
Intragenic Exonic |