Assay Details
Target Gene Details
Entrez Gene ID: | 19266 |
Gene Name: | protein tyrosine phosphatase, receptor type, D |
Gene Aliases: |
1110002J03Rik, 3000002J10Rik, B230219D21Rik, R-PTP-delta |
Location: |
Chr.4:75941237-78211895 on Build GRCm38 |
Assay Gene Location: | Within Exon 40 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprd | NM_011211.3 | 40 | 6602 | NP_035341.2 |
AK033873.1 | 3 | 1480 | BAC28503.1 | |
AK034067.1 | 3 | 1438 | BAC28569.1 | |
AK043201.1 | 5 | 1775 | BAC31488.1 | |
BC141409.1 | 30 | 5781 |
More Information
Set Membership: |
Intragenic Exonic |