Assay Details
Target Gene Details
Entrez Gene ID: | 76238 |
Gene Name: | glyoxylate reductase/hydroxypyruvate reductase |
Gene Aliases: |
1110059D05Rik, 6430629L09Rik |
Location: |
Chr.4:44981387-44992085 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grhpr | NM_080289.1 | 9 | 1119 | NP_525028.1 |
AK004307.1 | 9 | 1135 | ||
AK018293.1 | 9 | 1134 | ||
AK168351.1 | 9 | 1116 | BAE40288.1 | |
AK172201.1 | 9 | 1087 | BAE42878.1 | |
AY113690.1 | 9 | 1093 | AAM52985.1 | |
BC010194.1 | 9 | 1119 | AAH10194.1 |
More Information
Set Membership: |
Intragenic Exonic |