Assay Details
Target Gene Details
Entrez Gene ID: | 70902 |
Gene Name: | lysophosphatidylcholine acyltransferase 2B |
Gene Aliases: |
4921521K07Rik, Aytl1b |
Location: |
Chr.5:107431549-107435039 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpcat2b | NM_027599.3 | 2 | 822 | NP_081875.1 |
AK014940.1 | 2 | 824 | BAB29630.1 | |
AK029548.1 | 2 | 824 | BAC26509.1 | |
BC139095.1 | 2 | 447 | ||
BC139096.1 | 2 | 447 | ||
BY714455.1 |
More Information
Set Membership: |
Intragenic Exonic |