Assay Details
Target Gene Details
Entrez Gene ID: | 14745 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
AI326300, Edg2, Gpcr26, Kdt2, lpA1, vzg-1 |
Location: |
Chr.4:58435252-58554027 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpar1 | NM_001290486.1 | 2 | 492 | NP_001277415.1 |
NM_010336.2 | 3 | 607 | NP_034466.2 | |
NM_172989.1 | 3 | 696 | NP_766577.1 | |
AK030330.1 | 3 | 660 | BAC26905.1 | |
BC025425.1 | 3 | 606 | AAH25425.1 | |
BY716867.1 | 3 | 655 | ||
U48235.1 | 2 | 457 | AAC53035.1 | |
U70622.1 | 3 | 648 | AAC52923.1 |
More Information
Set Membership: |
Intragenic Exonic |