Assay Details
Target Gene Details
Entrez Gene ID: | 230787 |
Gene Name: | thymocyte selection associated family member 2 |
Gene Aliases: |
ICB-1 |
Location: |
Chr.4:132781837-132796364 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Themis2 | NM_001033308.2 | 6 | 2324 | NP_001028480.1 |
AK137465.1 | 5 | 4013 | ||
AK149775.1 | 6 | 2328 | ||
AK150471.1 | 6 | 2325 | BAE29588.1 | |
AK153536.1 | 6 | 2325 | BAE32073.1 | |
AK155536.1 | 6 | 2344 | BAE33313.1 | |
AK171009.1 | 6 | 2329 | BAE42180.1 | |
AK171858.1 | 6 | 2340 | ||
BC013712.1 | 6 | 2318 | AAH13712.1 |