Assay Details
Target Gene Details
Entrez Gene ID: | 20753 |
Gene Name: | small proline-rich protein 1A |
Gene Aliases: |
AI528815, SPR1a, mSPRR1A |
Location: |
Chr.3:92483954-92485881 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sprr1a | NM_009264.2 | 2 | 665 | NP_033290.1 |
AK144782.1 | 2 | 679 | BAE26064.1 | |
BC027534.1 | 2 | 665 | AAH27534.1 | |
X91824.1 | 2 | 658 | CAA62933.1 |