Assay Details
Target Gene Details
Entrez Gene ID: | 12349 |
Gene Name: | carbonic anhydrase 2 |
Gene Aliases: |
AI131712, CAII, Ca2, Car-2, Ltw-5, Lvtw-5 |
Location: |
Chr.3:14886278-14900770 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car2 | NM_009801.4 | 7 | 1260 | NP_033931.4 |
AK002333.1 | 7 | 995 | BAB22019.1 | |
AK002498.1 | 6 | 729 | BAB22146.2 | |
AK146015.1 | 7 | 997 | ||
BC055291.1 | 7 | 980 | AAH55291.1 | |
CD774528.1 | 1 | 548 | ||
K00811.1 | 7 | 961 | AAA37356.1 | |
M25944.1 | 1 | 213 | AAA39505.1 | |
U37706.1 | 1 | 156 |