Assay Details
Target Gene Details
Entrez Gene ID: | 229550 |
Gene Name: | RIKEN cDNA 9130204L05 gene |
Gene Aliases: |
Gm127 |
Location: |
Chr.3:91088137-91107429 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
9130204L05Rik | NM_001101461.2 | 3 | 385 | NP_001094931.1 |
AK078932.1 | 3 | 386 |
More Information
Set Membership: |
Intragenic Exonic |