Assay Details
Target Gene Details
Entrez Gene ID: | 229357 |
Gene Name: | G protein-coupled receptor 149 |
Gene Aliases: |
9630018L10Rik, Ieda, PGR10, R35 |
Location: |
Chr.3:62529963-62605442 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr149 | NM_177346.4 | 3 | 2113 | NP_796320.2 |
AK035931.1 | 3 | 676 | BAC29247.1 | |
AK136826.1 | 3 | 2114 | BAE23138.1 | |
BC119599.2 | 3 | 777 |
More Information
Set Membership: |
Intragenic Exonic |