Assay Details
Target Gene Details
Entrez Gene ID: | 26377 |
Gene Name: | dual adaptor for phosphotyrosine and 3-phosphoinositides 1 |
Gene Aliases: |
Bam32 |
Location: |
Chr.3:137930998-137981576 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dapp1 | NM_011932.2 | 9 | 2353 | NP_036062.1 |
AF186023.1 | 9 | 2328 | AAF14579.1 | |
AK145745.1 | 9 | 2319 | BAE26622.1 | |
AK169564.1 | 1 | 52 | ||
AK169584.1 | 8 | 2218 | BAE41242.1 |