Assay Details
Target Gene Details
Entrez Gene ID: | 12038 |
Gene Name: | butyrylcholinesterase |
Gene Aliases: |
C730038G20Rik |
Location: |
Chr.3:73635805-73708437 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bche | NM_009738.3 | 4 | 2686 | NP_033868.3 |
AK050337.1 | 4 | 2687 | BAC34196.1 | |
BC099977.1 | 4 | 2513 | AAH99977.1 |
More Information
Set Membership: |
Intragenic Exonic |