Assay Details
Target Gene Details
Entrez Gene ID: | 114889 |
Gene Name: | visual system homeobox 1 homolog (zebrafish) |
Gene Aliases: |
CHX10-like |
Location: |
Chr.2:150680702-150689137 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vsx1 | NM_054068.2 | 1 | 106 | NP_473409.1 |
AF395732.1 | 1 | 58 | AAL08947.1 | |
AK044393.1 | ||||
AK044687.1 | ||||
AK157311.1 | 1 | 329 | BAE34043.1 | |
BC057647.1 | 1 | 106 | AAH57647.1 | |
BC138456.1 | ||||
BC138460.1 |
More Information
Set Membership: |
Intragenic Exonic |