Assay Details
Target Gene Details
Entrez Gene ID: | 277432 |
Gene Name: | V-set and transmembrane domain containing 2-like |
Gene Aliases: |
Gm691 |
Location: |
Chr.2:157914653-157944719 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vstm2l | NM_198627.2 | 4 | 640 | NP_941029.2 |
BC058538.1 | AAH58538.1 | |||
BQ746346.1 | 3 | 357 |
More Information
Set Membership: |
Intragenic Exonic |