Assay Details
Target Gene Details
Entrez Gene ID: | 22325 |
Gene Name: | vav 2 oncogene |
Gene Aliases: |
2810040F13Rik, AI847175 |
Location: |
Chr.2:27262104-27427608 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vav2 | NM_009500.1 | 28 | 3204 | NP_033526.1 |
BC053060.1 | 30 | 3281 | AAH53060.1 | |
U37017.1 | 28 | 3204 | AAC52761.1 |