Assay Details
Target Gene Details
Entrez Gene ID: | 72033 |
Gene Name: | TSC22 domain family, member 2 |
Gene Aliases: |
1810043J12Rik, 5530402M19Rik |
Location: |
Chr.3:58415531-58466787 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsc22d2 | NM_001081229.1 | 4 | 7698 | NP_001074698.1 |
AK017449.1 | 1 | 633 | ||
AK141192.1 | 1 | 2718 | ||
AK170983.1 | 1 | 1180 |