Assay Details
Target Gene Details
Entrez Gene ID: | 18741 |
Gene Name: | paired-like homeodomain transcription factor 2 |
Gene Aliases: |
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg |
Location: |
Chr.3:129199878-129219594 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pitx2 | NM_001042502.2 | 3 | 1679 | NP_001035967.1 |
NM_001042504.2 | 4 | 1170 | NP_001035969.1 | |
NM_001286942.1 | 5 | 1269 | NP_001273871.1 | |
NM_001287048.1 | 3 | 1679 | NP_001273977.1 | |
NM_011098.4 | 5 | 1308 | NP_035228.2 | |
AB006320.1 | 3 | 1232 | BAA75247.1 | |
AB006321.1 | 5 | 1268 | BAA75248.1 | |
AF048723.1 | 4 | 1107 | AAC40086.1 | |
AF048724.1 | 3 | 1640 | AAC40087.1 | |
AF201091.1 | 3 | 1006 | AAF44618.1 | |
BC075660.1 | 3 | 1344 | AAH75660.1 | |
U70132.1 | 2 | 829 | AAB38864.1 | |
U80010.1 | 5 | 1243 | AAC53119.1 | |
U80011.1 | 4 | 1123 | AAC53120.1 | |
U80036.1 | 4 | 1141 | AAB38505.1 |
More Information
Set Membership: |
Intragenic Exonic |