Assay Details
Target Gene Details
Entrez Gene ID: | 545554 |
Gene Name: | ankyrin repeat domain 34A |
Gene Aliases: |
AW060394, Ankrd34 |
Location: |
Chr.3:96594826-96600126 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd34a | NM_001024851.3 | 2 | 978 | NP_001020022.2 |
BC052924.1 | 2 | 957 | AAH52924.1 | |
BC147489.1 | 1 | 771 | ||
BC150865.1 | 1 | 771 |