Assay Details
Target Gene Details
Entrez Gene ID: | 72175 |
Gene Name: | major facilitator superfamily domain containing 8 |
Gene Aliases: |
2810423E13Rik, AI836898, AV142426, Cln7 |
Location: |
Chr.3:40818112-40846856 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfsd8 | NM_028140.4 | 12 | 2273 | NP_082416.2 |
AK035291.1 | 12 | 2237 | ||
AK077957.1 | 12 | 2274 | BAC37083.1 | |
BC010483.1 | 12 | 2122 | AAH10483.1 |