Assay Details
Target Gene Details
Entrez Gene ID: | 22348 |
Gene Name: | solute carrier family 32 (GABA vesicular transporter), member 1 |
Gene Aliases: |
R75019, VGAT, Viaat |
Location: |
Chr.2:158610758-158615747 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc32a1 | NM_009508.2 | 2 | 1512 | NP_033534.2 |
AJ001598.1 | 2 | 1069 | CAA04864.1 | |
AK140468.1 | 2 | 1273 | BAE24402.1 | |
AK163661.1 | 2 | 1262 | BAE37446.1 | |
AK163754.1 | 2 | 1288 | BAE37482.1 | |
AK163929.1 | 1 | 371 | BAE37540.1 | |
AY578289.1 | 2 | 1512 | ||
BC052020.1 | 2 | 1503 | AAH52020.1 |