Assay Details
Target Gene Details
Entrez Gene ID: | 28030 |
Gene Name: | G elongation factor, mitochondrial 1 |
Gene Aliases: |
AW545374, D3Wsu133e, Gfm |
Location: |
Chr.3:67430115-67475068 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfm1 | NM_138591.2 | NP_613057.2 | ||
AF315511.1 | AAK58878.1 | |||
AK033214.1 | ||||
BC013093.1 | AAH13093.1 | |||
BC031772.1 | AAH31772.1 |
Target Gene Details
Entrez Gene ID: | 17035 |
Gene Name: | latexin |
Gene Aliases: |
- |
Location: |
Chr.3:67457999-67463907 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lxn | NM_016753.4 | 6 | 878 | NP_058033.2 |
AK032170.1 | 6 | 879 | BAC27739.1 | |
AK149981.1 | 6 | 838 | BAE29211.1 | |
BC115521.1 | 6 | 874 | ||
BC115522.1 | 6 | 876 | ||
D88769.1 | 6 | 784 | BAA13700.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |