Assay Details
Target Gene Details
Entrez Gene ID: | 21755 |
Gene Name: | protease, serine 39 |
Gene Aliases: |
Tesp1 |
Location: |
Chr.1:34498430-34503062 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss39 | NM_009355.2 | 5 | 876 | NP_033381.1 |
AB008910.1 | 5 | 876 | BAA26132.1 | |
BC049616.1 | 5 | 871 | ||
BC115665.2 | 5 | 875 | ||
BC115666.2 | 5 | 875 |
More Information
Set Membership: |
Intragenic Exonic |