Assay Details
Target Gene Details
Entrez Gene ID: | 14950 |
Gene Name: | histocompatibility 13 |
Gene Aliases: |
1200006O09Rik, 4930443L17Rik, 5031424B04Rik, AV020344, H-13, Hm13, PSL3, Spp |
Location: |
Chr.2:152669437-152708668 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H13 | NM_001159551.1 | 13 | 2027 | NP_001153023.1 |
NM_010376.4 | 12 | 1827 | NP_034506.1 | |
AK077983.1 | 5 | 922 | ||
AK080966.1 | 12 | 1789 | BAC38098.1 | |
AK131644.1 | 1 | 822 | ||
AK171790.1 | 11 | 1630 |
More Information
Set Membership: |
Intragenic Exonic |