Assay Details
Target Gene Details
Entrez Gene ID: | 227695 |
Gene Name: | DNA segment, Chr 2, Wayne State University 81, expressed |
Gene Aliases: |
9930028P05, AL033358 |
Location: |
Chr.2:30173447-30178495 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
D2Wsu81e | NM_172660.4 | 12 | 1459 | NP_766248.3 |
AK036945.1 | 10 | 1753 | ||
AK044983.1 | 10 | 2053 | ||
AK046148.1 | 11 | 1654 | BAC32613.1 | |
AK147162.1 | 12 | 1449 | BAE27727.1 | |
BC078441.1 | 9 | 2041 | AAH78441.1 | |
BY462424.1 | 1 | 51 |
Target Gene Details
Entrez Gene ID: | 13804 |
Gene Name: | endonuclease G |
Gene Aliases: |
- |
Location: |
Chr.2:30171401-30174069 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Endog | NM_007931.1 | 3 | 789 | NP_031957.1 |
AB012108.1 | 3 | 789 | BAA28168.1 | |
AK144459.1 | 3 | 821 | BAE25900.1 | |
BC030177.1 | 3 | 798 | AAH30177.1 | |
X99394.1 | CAA67768.1 |
More Information
Set Membership: |
Intragenic Exonic |