Assay Details
Target Gene Details
Entrez Gene ID: | 70239 |
Gene Name: | general transcription factor IIIC, polypeptide 5 |
Gene Aliases: |
2700084A09Rik, TFIIIC63, TFiiiC2-63 |
Location: |
Chr.2:28566245-28583746 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf3c5 | NM_001290484.1 | 1 | 79 | NP_001277413.1 |
NM_148928.3 | 1 | 79 | NP_683730.1 | |
AK137359.1 | 1 | 68 | ||
AK147014.1 | 1 | 57 | BAE27608.1 | |
BC027055.1 | 1 | 46 | AAH27055.1 | |
BC027247.1 | AAH27247.1 | |||
BC039171.1 | AAH39171.1 | |||
BQ179286.1 | 1 | 79 |
More Information
Set Membership: |
Intragenic Exonic |