Assay Details
Target Gene Details
Entrez Gene ID: | 58521 |
Gene Name: | EP300 interacting inhibitor of differentiation 1 |
Gene Aliases: |
2610002K20Rik, Cri1, EID-1, ORF12, PNAS-22, PTD014 |
Location: |
Chr.2:125673100-125675643 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eid1 | NM_025613.3 | 1 | 427 | NP_079889.2 |
AF509968.1 | 1 | 365 | AAM34760.1 | |
AK002559.1 | 1 | 433 | BAB22187.1 | |
AK005258.1 | 1 | 397 | BAB23911.1 | |
AK011293.1 | 1 | 419 | BAB27521.1 | |
AK012453.1 | 1 | 418 | BAB28248.1 | |
AK014440.1 | 1 | 373 | BAB29352.1 | |
AK017543.1 | 1 | 401 | BAB30797.1 | |
AK077783.1 | 1 | 407 | BAC37007.1 | |
AK078402.1 | 1 | 373 | BAC37257.1 | |
AK078409.1 | 1 | 373 | BAC37261.1 | |
AK078826.1 | 1 | 351 | BAC37412.1 | |
AK148555.1 | 1 | 368 | BAE28618.1 | |
AK169604.1 | 1 | 401 | BAE41254.1 | |
AK172612.1 | 1 | 208 | BAE43093.1 | |
AL357453.1 | 1 | 393 | ||
BY703074.1 | 1 | 426 |
Target Gene Details
Entrez Gene ID: | 271849 |
Gene Name: | SHC (Src homology 2 domain containing) family, member 4 |
Gene Aliases: |
6230417E10Rik, 9930029B02Rik, Gm685, RaLP |
Location: |
Chr.2:125627447-125724160 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shc4 | NM_199022.2 | NP_950187.1 | ||
AY464564.1 | AAR19362.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |