Assay Details
Target Gene Details
Entrez Gene ID: | 18286 |
Gene Name: | outer dense fiber of sperm tails 2 |
Gene Aliases: |
AI848335, MMTEST29 |
Location: |
Chr.2:29889720-29931746 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Odf2 | NM_001113213.1 | NP_001106684.1 | ||
NM_001113214.1 | 17 | 2339 | NP_001106685.1 | |
NM_001177659.1 | NP_001171130.1 | |||
NM_001177661.1 | 16 | 2185 | NP_001171132.1 | |
NM_013615.3 | 17 | 2312 | NP_038643.1 | |
AF000968.1 | 15 | 2159 | AAB65157.1 | |
AF034105.1 | 17 | 2275 | AAB87525.1 | |
AK143726.1 | BAE25517.1 | |||
AK165588.1 | ||||
AK168415.1 | 17 | 2241 | BAE40330.1 | |
BC057001.1 | AAH57001.1 |