Assay Details
Target Gene Details
Entrez Gene ID: | 14683 |
Gene Name: | GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus |
Gene Aliases: |
5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH |
Location: |
Chr.2:174283444-174346744 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnas | NM_001077507.1 | 1 | 395 | NP_001070975.1 |
NM_010309.3 | 1 | 395 | NP_034439.2 | |
NM_019690.2 | NP_062664.2 | |||
NM_022000.2 | NP_068840.2 | |||
NM_201617.1 | 1 | 395 | NP_963911.1 | |
NM_201618.1 | 1 | 395 | NP_963912.1 | |
AF107847.1 | AAD11805.1 | |||
AF107848.1 | AAD11806.1 | |||
AF175305.1 | AAD55061.1 | |||
AJ010163.1 | CAA09026.1 | |||
AY519501.1 | 1 | 395 | AAS00601.1 | |
AY519502.1 | 1 | 395 | AAS00602.1 | |
AY519503.1 | 1 | 395 | AAS00603.1 | |
AY519504.1 | 1 | 395 | AAS00604.1 | |
BY718446.1 |
More Information
Set Membership: |
Intragenic Exonic |